missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA B7 Antibody (SR1670), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | HLA B7 |
|---|---|
| Clone | SR1670 |
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18614065
|
Novus Biologicals
NBP3-22213-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18678884
|
Novus Biologicals
NBP3-22213-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HLA B7 Monoclonal antibody specifically detects HLA B7 in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
| HLA B7 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 3106 | |
| IgG | |
| Affinity purified |
| SR1670 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| ankylosing spondylitis, AS, b'DT, Bw-22, Bw-4, Bw-41, Bw-44, Bw-45, Bw-46, Bw-47, Bw-48, Bw-50, Bw-52, Bw-53, Bw-54, Bw-55, Bw-56, Bw-57, Bw-58, Bw-60, HLA class I histocompatibility antigen, B alpha chain, HLA class I histocompatibility antigen, B-12 alpha chain, HLA class I histocompatibility antigen, B-21 alpha chain, HLA class I histocompatibility antigen, B-5 alpha chain, HLA class I histocompatibility antigen, B-7 alpha chain, HLAB, HLA-B27, HLA-B73, leukocyte antigen class I-B, lymphocyte antigen, major histocompatibility complex, class I, B, MGC111087, MHC class I antigen B*13, MHC class I antigen B*14, MHC class I antigen B*15, MHC class I antigen B*18, MHC class I antigen B*27, MHC class I antigen B*35, MHC class I antigen B*37, MHC class I antigen B*38, MHC class I antigen B*39, MHC class I antigen B*40, MHC class I antigen B*41, MHC class I antigen B*42, MHC class I antigen B*44, MHC class I antigen B*45, MHC class I antigen B*46, MHC class I antigen B*47, MHC class I anti | |
| A synthesized peptide derived from human HLA B7 (Uniprot #: P01889) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title