missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-B7 Mouse anti-Human, Brilliant Violet 510, Clone: BB7.1, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 747886
This item is not returnable.
View return policy
Description
HLA-B7 Monoclonal antibody specifically detects HLA-B7 in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA-B7 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Mouse | |
| RUO | |
| HLA-B7 | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| BB7.1 | |
| Brilliant Violet 510 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction