Learn More
HLA-C Mouse anti-Human, PE, Clone: DT-9 (also known as DT9), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 566372
Description
The DT-9 monoclonal antibody specifically recognizes Human Leukocyte Antigen (HLA-C), a polymorphic major histocompatibility complex (MHC) class I antigen. HLA-C is a heterodimer comprised of the HLA-C alpha chain, a ñ45 kDa type I transmembrane glycoprotein, and a ñ12 kDa Beta-2 (β2)-microglobulin light chain. HLA-C is expressed on nearly all cells, and plays a role in the antigen-specific, MHC-restricted presentation of small peptides to CD8+ T cells in the generation of immunity or tolerance. HLA-C may also bind to regulatory MHC class I antigen-selective receptors expressed by CD8+ T cells and natural killer (NK) cells. The DT-9 antibody also recognizes some rare HLA-A and HLA-B allotypes and the nonclassical HLA-E molecule.
R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
Specifications
| HLA-C | |
| DT-9 (also known as DT9) | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
| Monoclonal | |
| PE | |
| HLAC, HLC-C, HLA-JY3, MHC class I antigen C, PSORS1, HLA-E | |
| Purified Human Tamarin MHC Class I Molecules | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.