Learn More
HLA class I ABC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15240-1-AP-150UL
 View more versions of this product
Product Code. 16844334
  • € 410.00 / 150µL
Description
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determits important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumour defence. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules.
Specifications
Specifications
HLA class I ABC
Polyclonal
Unconjugated
HLA-A
HLA A, HLA class I, HLA class I (HLA-A), HLA class I ABC, HLA-A, MHC class I antigen A*3
Rabbit
Antigen Affinity Chromatography
RUO
3105
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation
0.3 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P30443
HLA-A
HLA class I ABC Fusion Protein Ag7370
150 μL
Primary
Human, Mouse