missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA Class I Antibody (MEM-81) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-412
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| HLA Class I | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry 1-5 ug/ml, CyTOF-ready | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody MEM-81 reacts with human classical MHC Class I molecules in native cell-surface forms. MHC Class I molecules (MHC Class Ia) are expressed on the surface of human nucleated cell types. | |
| Store at 4C. Do not freeze. |
| Flow Cytometry, CyTOF | |
| MEM-81 | |
| Unconjugated | |
| P04439 | |
| HLA-A | |
| Leukocytes of a patient suffering with LGL type leukemia. | |
| 0.1 mg | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction