missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA Class I Antibody (W6/32) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7098JF585
This item is not returnable.
View return policy
Description
HLA Class I Monoclonal antibody specifically detects HLA Class I in Human samples. It is validated for Flow Cytometry,CyTOF-reported
Specifications
| HLA Class I | |
| Monoclonal | |
| Janelia Fluor 585 | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A&1, MHC class I antigen A&11, MHC class I antigen A&80 | |
| Membranes from human tonsillar lymphocytes, | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| W6/32 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction