Learn More
HLA DMA Antibody (5C9NB), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-44300AF594
Product Code. 18171508
  • € 456.00 / 0.10mL
Description
HLA DMA Monoclonal specifically detects HLA DMA in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA DMA
Monoclonal
Alexa Fluor 594
50 mM sodium borate with 0.05% sodium azide
HLA-DMA
Partial recombinant human HLA DMA protein (between amino acids 10-300) [Entrez NP_006111]
Primary
Human
IgG1 κ
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
5C9NB
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Cell Surface)
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
Mouse
0.1 mL
3108
Store at 4°C in the dark.