missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DOB Antibody (DOB 10E6/5), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50420MFV450
This item is not returnable.
View return policy
Description
HLA DOB Monoclonal antibody specifically detects HLA DOB in Human samples. It is validated for Western Blot, ELISA
Specifications
| HLA DOB | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DOB 10E6/5 | |
| Western Blot, ELISA | |
| DOB, FLJ57033, HLA class II histocompatibility antigen, DO beta chain, major histocompatibility complex, class II, DO beta, MHC class II antigen DOB | |
| C-terminal peptide MHC class II | |
| 0.1 mL | |
| Asthma, Immunology | |
| 3112 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction