missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DPA Antibody (TAL3C3), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50601CL1
This item is not returnable.
View return policy
Description
HLA DPA Monoclonal antibody specifically detects HLA DPA in Human samples. It is validated for Immunohistochemistry,Western Blot,Radioimmune Assays
Specifications
| HLA DPA | |
| Monoclonal | |
| CoraFluor 1 | |
| DP alpha 1 chain, HLA class II histocompatibility antigen, DP alpha chain, HLADP, HLA-SB alpha chain, major histocompatibility complex, class II, DP alpha 1, MHC class II antigen, MHC class II DP3-alpha, MHC class II DPA1, MHC class II HLA-DPA1 antigen, Primed lymphocyte test-1 | |
| Bristol 8 separated alpha chain preparation | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Immunology | |
| 3113 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Radioimmune Assays (RIA) | |
| TAL3C3 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction