missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DPA Antibody (TAL3C3), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50601MFV610
This item is not returnable.
View return policy
Description
HLA DPA Monoclonal antibody specifically detects HLA DPA in Human samples. It is validated for Western Blot, Immunohistochemistry, Radioimmune Assays
Specifications
| HLA DPA | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Radioimmune Assays (RIA) | |
| TAL3C3 | |
| Western Blot, Immunohistochemistry, Radioimmunoassay | |
| DP alpha 1 chain, HLA class II histocompatibility antigen, DP alpha chain, HLADP, HLA-SB alpha chain, major histocompatibility complex, class II, DP alpha 1, MHC class II antigen, MHC class II DP3-alpha, MHC class II DPA1, MHC class II HLA-DPA1 antigen, Primed lymphocyte test-1 | |
| Bristol 8 separated alpha chain preparation | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Immunology | |
| 3113 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction