missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DPB1 Antibody (SPM421), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11503IR
This item is not returnable.
View return policy
Description
HLA DPB1 Monoclonal antibody specifically detects HLA DPB1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence
Specifications
| HLA DPB1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunofluorescence | |
| SPM421 | |
| Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| DP beta 1 chain, DPB1, HLA class II histocompatibility antigen, DP(W4) beta chain, HLA DP14-beta chain, HLA-DP histocompatibility type, beta-1 subunit, HLA-DP1B, HLA-DPB, major histocompatibility complex class II HLA DPB1 protein, major histocompatibility complex, class II, DP beta 1, MHC class II antigen beta chain, MHC class II antigen DP beta 1 chain, MHC class II antigen DPB1, MHC class II antigen DPbeta1, MHC class II HLA-DP-beta, MHC HLA DPB1 | |
| Non-T, non-B human acute lymphoblastic leukemia REH6 cell line | |
| 0.1 mL | |
| Asthma, Diabetes Research, Immunology | |
| 3115 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction