missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-DPB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA527780
This item is not returnable.
View return policy
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HLA-DPB belongs to the HLA class II beta chain paralogues. This class II molecule is a heterodimer consisting of an alpha (DPA) and a beta chain (DPB), both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The beta chain is approximately 26-28 kDa and its gene contains 6 exons. Exon one encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, exon 4 encodes the transmembrane domain and exon 5 encodes the cytoplasmic tail. Within the DP molecule both the alpha chain and the beta chain contain the polymorphisms specifying the peptide binding specificities, resulting in up to 4 different molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DPB1 | |
| Polyclonal | |
| Unconjugated | |
| HLA-DPB1 | |
| beta1 domain MHC class II HLA DPB; CD; CELIAC1; class II HLA beta chain; DC-1 alpha chain; DC-alpha; DPB1; DQ-A1; GSE; histocompatibility antigen HLA-DR alpha; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DQ alpha 1 chain; HLA class II histocompatibility antigen, DQ(W3) alpha chain; HLA class II histocompatibility antigen, DR alpha chain; HLA DP14-beta chain; HLA-DCA; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DQA; HLA-DQA1; HLA-DRA; HLA-DRA1; leucocyte antigen DQA1; leukocyte antigen alpha chain; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DQ alpha 1; major histocompatibility complex, class II, DR alpha; MHC cell surface glycoprotein; MHC cla; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DRA; MHC class II DQA1; MHC class II HLA-D alpha glycoprotein; MHC class II HLA-DP-beta-1; MHC class II HLA-DQ-alpha-1; MHC class II HLA-DRB1; MHC class II surface glycoprotein; MHC HLA DPB1; MHC HLA-DQ alpha; MLRW | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 3115 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P04440 | |
| HLA-DPB1 | |
| Recombinant protein encompassing a sequence within the center region of human HLA-DPB1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction