Learn More
HLA DQ Antibody (B-K27) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-18113JF669
Product Code. 30165038
  • ONLINE EXCLUSIVE PRICE
    € 421.00
    € 397.95 / 0.10mL
    Save € 23.05
    5% Off
Description
HLA DQ Monoclonal antibody specifically detects HLA DQ in Human samples. It is validated for Flow Cytometry,Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA DQ
Monoclonal
Janelia Fluor 669
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
T4+ CTL clone HG-38. Hybridoma: Myeloma X63/AG.8653 x Balb/c spleen cells
0.1 mL
Adaptive Immunity, Asthma, Immunology
3117
Store at 4°C in the dark.
IgG2a κ
Flow Cytometry, Immunoprecipitation
B-K27
50mM Sodium Borate
Mouse
Affinity purified