Learn More
HLA DQ Antibody (K274.3G8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-11305-0.025mg
 View more versions of this product
Product Code. 18666797
  • € 188.00 / 25µg
Description
HLA DQ Monoclonal antibody specifically detects HLA DQ in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA DQ
Monoclonal
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Pig
Purified
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
K274.3G8
Flow Cytometry 1:10 - 1:100, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Porcine peripheral blood lymphocytes
0.025 mg
Adaptive Immunity, Asthma, Immunology
3117
Store at 4°C.