missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ Antibody (TAL 4.1), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2700AF750
This item is not returnable.
View return policy
Description
HLA DQ Monoclonal specifically detects HLA DQ in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.
Specifications
| HLA DQ | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HLA-DQA1 | |
| Whole cells from the B line WT46 | |
| 0.1 mL | |
| 3117 | |
| Human | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| TAL 4.1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Mouse | |
| Protein A purified | |
| Primary | |
| Human Leukocyte Antigen DQ1 (includes DQ5 and 6) | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction