missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ Antibody (TAL 4.1), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2700JF646
This item is not returnable.
View return policy
Description
HLA DQ Monoclonal specifically detects HLA DQ in Human samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| HLADQ | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HLA-DQA1 | |
| Whole cells from the B line WT46 | |
| 0.1 mL | |
| Primary | |
| Human Leukocyte Antigen DQ1 (includes DQ5 and 6) | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| TAL 4.1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Asthma, Immunology | |
| 3117 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction