missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ/DR/DP Antibody (Bu26), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-64162CL1
This item is not returnable.
View return policy
Description
HLA DQ/DR/DP Monoclonal antibody specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry
Specifications
| HLA DQ/DR/DP | |
| Monoclonal | |
| CoraFluor 1 | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Made to Human HLA DP DQ DR | |
| 0.1 mL | |
| Adaptive Immunity, Asthma, Diabetes Research, Immunology | |
| 3117 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| Bu26 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction