Learn More
HLA DQ/DR/DP Antibody (Bu26), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-64162UV
Product Code. 18035895
  • € 608.00 / 0.10mL
Description
HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA DQ/DR/DP
Monoclonal
DyLight 350
P01909
HLA-DQA1
Made to Human HLA DP DQ DR
0.1 mL
Adaptive Immunity, Asthma, Immunology
3117
Human
Purified
Flow Cytometry
Bu26
Flow Cytometry
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Mouse
Protein G purified
RUO
Primary
NB100-64162 recognizes the beta chains of class II human major histocompatibility (MHC) antigens. The antibody reacts with DP, DQ, DR beta chains, which are expressed by antigen presenting cells, B- cells, monocytes and activated T lymphocytes.
Store at 4C in the dark.