missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ/DR/DP Mouse anti-Human, Clone: rHLA-Pan/3475, Novus Biologicals™
Mouse Monoclonal Antibody
€ 522.00
Specifications
| Antigen | HLA DQ/DR/DP |
|---|---|
| Clone | rHLA-Pan/3475 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1 to 2 μg/million cells, ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
HLA DQ/DR/DP Monoclonal specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HLA DQ/DR/DP | |
| 0.2 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Adaptive Immunity, Asthma, Diabetes Research, Immunology | |
| 10 mM PBS with 0.05% BSA | |
| 3117 | |
| IgG1 κ | |
| Protein A or G purified |
| rHLA-Pan/3475 | |
| Flow Cytometry 1 to 2 μg/million cells, ELISA 1-5 μg/mL, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Non-T, non-B human acute lymphoblastic leukemia REH6 cells | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title