Learn More
HLA-DQ Mouse anti-Human, Brilliant Violet 711, Clone: Tu169, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 742612
Description
The TU169 monoclonal antibody specifically binds to the human Major Histocompatibility Complex (MHC) Class II HLA-DQ1 and DQ2 antigens and weakly to HLA-DQ3. HLA-DQ antigens exits as heterodimers comprised of polymorphic transmembrane HLA-DQ alpha and beta glycoproteins. HLA-DQ is expressed primarily on B cells, monocytes, macrophages, dendritic cells and activated T lymphocytes. HLA-DQ is involved in presenting peptidic antigens to CD4+ T lymphocytes. This antibody does not cross-react with HLA-DR2 and HLA-DR7. This antibody fixes complement.
The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
Specifications
| HLA-DQ | |
| Tu169 | |
| Brilliant Violet 711 | |
| MHC class II HLA-DQ, MHC HLA-DQ | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.