missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DQ Mouse anti-Human, Oligo, Clone: Tu169, BD™ AbSeq ™
Mouse Monoclonal Antibody
Brand: BD Biosciences 940233
This item is not returnable.
View return policy
Description
HLA-DQ Monoclonal antibody specifically detects HLA-DQ in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
| HLA-DQ | |
| Monoclonal | |
| Oligo | |
| MHC class II HLA-DQ; MHC HLA-DQ | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
| Single Cell 3' Sequencing | |
| Clone Tu169 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction