missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DQ1/DQ3 Antibody (HL-37), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-405AF594
This item is not returnable.
View return policy
Description
HLA DQ1/DQ3 Monoclonal specifically detects HLA DQ1/DQ3 in Human, Porcine samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| HLA DQ/DQ3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HLA-DQA1 | |
| Burkitt′s lymphoma cell line Raj | |
| Primary | |
| The antibody HL-37 reacts with polymorphic determinant on human HLA-DQ1 and HLA-DQ3 molecules (recognized epitope was found on isolated beta chain of DQ1), but does not react with HLA-DQ2. DQ is the isotype of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages). | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| HL-37 | |
| Western Blot, Flow Cytometry | |
| CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha | |
| Mouse | |
| 0.1 mL | |
| 3117 | |
| Human, Pig | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction