missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-DQA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116333
This item is not returnable.
View return policy
Description
HLA-DQA2 Polyclonal Antibody for Western Blot
HLA-DQA2 belongs to the HLA class II alpha chain family. The encoded protein forms a heterodimer with a class II beta chain. It is located in intracellular vesicles and plays a central role in the peptide loading of MHC class II molecules by helping to release the CLIP molecule from the peptide binding site. Class II molecules are expressed in antigen presenting cells (B lymphocytes, dendritic cells, macrophages) and are used to present antigenic peptides on the cell surface to be recognized by CD4 T-cells. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DQA2 | |
| Polyclonal | |
| Unconjugated | |
| HLA-DQA2 | |
| DQ alpha 2; DX ALPHA; DX alpha chain; DX-ALPHA; HLA class II histocompatibility antigen, DQ alpha 2 chain; HLA class II histocompatibility antigen, DQ(6) alpha chain; HLA DXA; HLA-DQA1; HLA-DQA2; HLA-DXA; major histocompatibility complex, class II, DQ alpha 2; MHC class II DQA2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 3118 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.35 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P01906 | |
| HLA-DQA2 | |
| KLH conjugated synthetic peptide between 39-66 amino acids from the Central region of human HLA-DQA2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction