missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-DQw1 Monoclonal Antibody (Genox 3.53)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA146297
This item is not returnable.
View return policy
Description
HLA-DQw1 Monoclonal Antibody for ICC/IF, IHC (F), Flow, IP, ELISA
Human Leukocyte Antigens are highly polymorphic proteins that are involved in the presentation of antigens to the T-cell receptor. There are two classes of HLA antigens, class I (HLA A, HLA B and HLA C) and class II (HLA D). This antibody can be used for HLA typing. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DQw1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P01920 | |
| HLA-DQB1 | |
| Preparation of human Bristol 8 glycoprotein from the Bristol 8 B lymphoblastoid cell line. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Immunocytochemistry | |
| Genox 3.53 | |
| Unconjugated | |
| HLA-DQB1 | |
| CELIAC1; HLA class II antigen; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1; lymphocyte antigen; major histocompatibility complex, class II, DQ beta 1; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II DQ beta chain; MHC class II HLA-DQ beta glycoprotein; MHC class2 antigen; MHC DQ beta | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 3119 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction