missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Antibody (37.68), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65424IR
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal specifically detects HLA-DR in Human, Bovine, Goat, Sheep samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| HLA-DR | |
| Monoclonal | |
| DyLight 755 | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-65424 recognizes a monomorphic epitope on ovine MHC class II DR molecules. The distribution of ovine DR molecules on T lymphocytes has been shown to vary with immune status and age. Initially T lymphocytes lack class II molecules, then T lymphocytes progressively express DR molecules. | |
| Store at 4C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 37.68 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| HLA-DRA | |
| Splenocytes from ATL mice (1) | |
| 0.125 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3122 | |
| Human, Bovine, Goat, Sheep | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction