missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Antibody (HLA-DRA/6839R), Janelia Fluor™ 646, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20834JF646
This item is not returnable.
View return policy
Description
HLA-DR Monoclonal antibody specifically detects HLA-DR in Human samples. It is validated for Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| HLA-DR | |
| Monoclonal | |
| Janelia Fluor 646 | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Recombinant fragment (around aa1-200) of human HLA-DR protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Adaptive Immunity, B Cell Development and Differentiation Markers, Cell Biology, Cellular Markers, Diabetes Research, Immunology, Major Histocompatibility Complexes | |
| 3122 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| HLA-DRA/6839R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction