missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR, DP, DQ Mouse anti-Human, BV750, Clone: Tu39 (also known as TÜ39), BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 746979
This item is not returnable.
View return policy
Description
HLA-DR, DP, DQ Monoclonal antibody specifically detects HLA-DR, DP, DQ in Human samples. It is validated for Flow Cytometry
Specifications
| HLA-DR, DP, DQ | |
| Monoclonal | |
| Brilliant Violet 750 | |
| Tü39; MHC class II HLA-DR, DP, DQ | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BV750 under optimal conditions that minimize unconjugated dye and antibody. |
| Flow Cytometry | |
| Tu39 (also known as TÜ39) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| RUO | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur