missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-DR Mouse, APC-Cy7, Clone: L243, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QAB86-APC7-100Tests
This item is not returnable.
View return policy
Specifications
| HLA-DR | |
| Monoclonal | |
| 5μL/Test | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| HLA-DRA | |
| Protein A/G Purified | |
| RUO | |
| 3122 | |
| IgG2a κ |
| Flow Cytometry | |
| L243 | |
| APC-Cyanine7 | |
| NP_061984 | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction