missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRA Antibody (19-26.1 (same as MB-26.1)), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-11518CL1
This item is not returnable.
View return policy
Description
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin),Immunofluorescence,Immunocytochemistry/Immunofluorescence
Specifications
| HLA DRA | |
| Monoclonal | |
| CoraFluor 1 | |
| FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
| Raji cells | |
| 0.1 mL | |
| Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
| 3122 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry/Immunofluorescence | |
| 19-26.1 (same as MB-26.1) | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction