missing translation for 'onlineSavingsMsg'
Learn More

HLA DRA Antibody (CL10964) [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30528784 Shop All Bio Techne Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
0.1 mL
missing translation for 'unitSize'
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30528784 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30528784 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP344200APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen HLA DRA
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL10964
Conjugate APC
Formulation PBS
Gene Alias FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P01903, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Biology, Diabetes Research, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3122
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.