missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRB1 Antibody (DA2), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2705PEATT594
This item is not returnable.
View return policy
Description
HLA DRB1 Monoclonal specifically detects HLA DRB1 in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA DRB1 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| HLA-DRB1 | |
| The membranes of LKT cells (a B lymphoblastoid cell line) | |
| 0.1 mL | |
| Primary | |
| DA2 will detect dimers and the beta chain by western blot. DA2 will bind all DR and DP antigens except DR7. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| DA2 | |
| Flow Cytometry | |
| Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
| Mouse | |
| Protein A purified | |
| Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology | |
| 3123.0 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction