missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA DRB1 Antibody (TDR 31.1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50421
This item is not returnable.
View return policy
Description
HLA DRB1 Monoclonal antibody specifically detects HLA DRB1 in Human samples. It is validated for Flow Cytometry, ELISA, Radioimmune Assays, CyTOF
Specifications
| HLA DRB1 | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000, CyTOF-ready | |
| Clone P2-beta-3, DR1, DR-1, DR12, DR-12, DR13, DR-13, DR14, DR-14, DR16, DR-16, DR4, DR-4, DR5, DR-5, DR7, DR-7, DR8, DR-8, DR9, DR-9, DRB1, DRw10, DRw11, DRw8, DW2.2/DR2.2, FLJ75017, FLJ76359, HLA class II antigen beta chain, HLA class II histocompatibility antigen, DR-1 beta chain, HLA-DR1B, HLA-DRB, HLA-DRB1*, HLA-DRB2, HLA-DR-beta 1, human leucocyte antigen DRB1, leucocyte antigen DR beta 1 chain, leucocyte antigen DRB1, lymphocyte antigen DRB1, major histocompatibility complex, class II, DR beta 1, MHC class II antigen DRB1*1, MHC class II antigen DRB1*10, MHC class II antigen DRB1*11, MHC class II antigen DRB1*12, MHC class II antigen DRB1*13, MHC class II antigen DRB1*14, MHC class II antigen DRB1*15, MHC class II antigen DRB1*16, MHC class II antigen DRB1*3, MHC class II antigen DRB1*4, MHC class II antigen DRB1*7, MHC class II antigen DRB1*8, MHC class II antigen DRB1*9, MHC class II antigen HLA-DR13, MHC class II HLA-DR beta 1 chain, MHC class II HLA-DR-beta cell surface gly | |
| Ia fraction from Jy human B lymphoblastoid cell line. Class II from JY human B lymphoblastoid cell line. | |
| 0.1 mL | |
| Asthma, Immunology | |
| 3123 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Flow Cytometry, ELISA, Radioimmune Assays (RIA), CyTOF | |
| TDR 31.1 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction