missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA G Antibody (87G), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43123B
This item is not returnable.
View return policy
Description
HLA G Monoclonal specifically detects HLA G in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| HLA G | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| HLA-G | |
| 721.221 cells transfected with full length HLA G cDNA with high surface HLA G expression. [UniProt# P17693] | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
| 3135 | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 87G | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC-G, sHLA-G | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Membrane bound HLA G1 and soluble HLA G5. | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction