Learn More
HLA G Antibody (HLAG/8393R), DyLight 650, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-20690C
Product Code. 18572799
  • € 726.00 / 0.10mL
Description
HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HLA G
Monoclonal
DyLight 650
b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G
Purified His-tagged HLA G protein.
0.1 mL
Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites
3135
Store at 4°C in the dark.
IgG κ
Immunohistochemistry (Paraffin)
HLAG/8393R
50 mM Sodium Borate
Rabbit
Protein A or G purified