missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA G Antibody (HLAG/8393R) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-20690JF585
This item is not returnable.
View return policy
Description
HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HLA G | |
| Monoclonal | |
| Janelia Fluor 585 | |
| b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G | |
| Purified His-tagged HLA G protein. | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
| 3135 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| HLAG/8393R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction