missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA G Antibody (MEM-G/4), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-533FR
This item is not returnable.
View return policy
Description
HLA G Monoclonal antibody specifically detects HLA G in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
| HLA G | |
| Monoclonal | |
| DyLight 680 | |
| b2 microglobulin, HLA class I histocompatibility antigen, alpha chain G, HLA class I molecule, HLA G antigen, HLA-6.0, HLAG, HLA-G, HLA-G histocompatibility antigen, class I, G, major histocompatibility complex, class I, G, MHC class I antigen G, MHC class Ib antigen, MHC-G, mutant MHC class I antigen, mutant MHC class Ib antigen, sHLA-G, Soluble HLA class I histocompatibility antigen, alpha chain G | |
| Recombinant human HLA G Antibody (MEM-G/4) denaturated heavy chain. | |
| 0.1 mL | |
| Adaptive Immunity, Autoimmune Diseases, Cancer, Developmental Biology, Immunology, Inflammation, Innate Immunity, Virology Bacteria and Parasites | |
| 3135 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF | |
| MEM-G/4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction