missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-G Mouse anti-Human, BV605, Clone: 87G, BD OptiBuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 751657
This item is not returnable.
View return policy
Description
HLA-G Monoclonal antibody specifically detects HLA-G in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA-G | |
| Monoclonal | |
| 0.2 mg/mL | |
| Hu HLA-G BV605 87G 50 μg | |
| HLA-G-transfected Cells | |
| RUO | |
| HLA-G | |
| Refrigerate (2 to 8°C) | |
| Human |
| Flow Cytometry | |
| 87G | |
| Brilliant Violet 605 | |
| Mouse | |
| 1 x 50 μg | |
| Primary | |
| Human | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction