Learn More
HLX Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544857
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: WFQNRRMKWR HSKEAQAQKD KDKEAGEKPS GGAPAADGEQ DERSPSRSEG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 75%; Zebrafish: 93%
HLX Polyclonal antibody specifically detects HLX in Human samples. It is validated for Western Blot.Specifications
HLX | |
Polyclonal | |
Unconjugated | |
HLX | |
dbx1a; developing brain homeobox 1a; developing brain homeobox protein 1-A; fc15c04; H2.0 homeobox 1; H2.0 like homeobox; H2.0-like homeo box 1; H2.0-like homeo box-1; H2.0-like homeobox; H2.0-like homeobox 1; H2.0-like homeobox protein; HB24; Hlx; hlx1; hlx-1; homeobox protein; homeobox protein DBX1-A; Homeobox protein HB24; homeobox protein HLX1; LOW QUALITY PROTEIN: H2.0-like homeobox protein; wu:fc15c04; zgc:109861 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3142 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q14774 | |
HLX | |
Synthetic peptide directed towards the middle region of human HLX | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |