Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Use Epredia™ HM 525 NX blade holders with HM 525 NX and other Epredia™ cryostats.
Brand:  Epredia™ 705720
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12023119
  • € 1465.00 / Each
Estimated Shipment: 06-09-2021
Add to basket
Description
Description
Standard Knife Carrier:
  • Allows for insertion of standard knives, with clearance angle adjustment
  • Can be moved horizontally on precision dovetail guide with quick clamp system
  • Includes anti-roll plate
Disposable Blade Carrier EC:
  • Allows for insertion of high- and low-profile blades, with clearance angle adjustment
  • Can be moved on precision dovetail guide with quick clamp plate
  • Includes anti-roll plate
Disposable Blade Carrier EV:
  • Use with vacutome system
  • Allows for insertion of high- and low-profile disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
  • Use with vacutome system
  • Allows for insertion of disposable blades
  • Includes a vacuum anti-roll hood for stretching and disposing of section waste
  • Features a slot that is opened and closed by a vacuum-controlled flap
Specifications
Specifications
Blade Holder
Microm™ HM 560 cryostats
Cryostat Blade Holders