Learn More
Epredia™ HM 525 NX Cryostat Blade Holders
Shop All Epredia Products
Click to view available options
Product Type:
Disposable Blade Carrier EC
Disposable Blade Carrier EC70
Disposable Blade Carrier EV
Standard Knife Carrier SM/SMV
Standard Knife Holder
For Use With (Equipment):
HM 525 NX and HM 550
Microm HM 560 Cryostat
Unit Size:
Each
Description
Standard Knife Carrier:
- Allows for insertion of standard knives, with clearance angle adjustment
- Can be moved horizontally on precision dovetail guide with quick clamp system
- Includes anti-roll plate
Disposable Blade Carrier EC:
- Allows for insertion of high- and low-profile blades, with clearance angle adjustment
- Can be moved on precision dovetail guide with quick clamp plate
- Includes anti-roll plate
Disposable Blade Carrier EV:
- Use with vacutome system
- Allows for insertion of high- and low-profile disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Magnetic Disposable Blade Carrier MV:
- Use with vacutome system
- Allows for insertion of disposable blades
- Includes a vacuum anti-roll hood for stretching and disposing of section waste
- Features a slot that is opened and closed by a vacuum-controlled flap
Specifications
Specifications
| Product Type | Standard Knife Holder |
| For Use With (Equipment) | HM 525 NX and HM 550 |