missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ HM 650V Vibrating Blade Microtome Specimen Stage F (Flat)for medium to large specimens or tissue (from 10mm to 45mm)

Use these Epredia™ HM 650V Vibrating Blade Microtome Accessories with the Epredia™ HM650V Vibrating Blade Microtome.

Brand:  Epredia™ 648200

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12003099

  • € 288.00 / Each


Estimated Shipment: 18-10-2021
Add to basket

Description

Description

Standard buffer tray:
  • Hold stages in place using a magnet
  • Fill tray with all water-based buffers and solutions
Cooling element for buffer solution:
  • Use pre-cooled in the standard buffer tray for cooling tissue
  • Use frozen together with buffer solution to prevent the frozen buffer solution from inflating
Large-field magnifier:
  • Magnification: 2.5x
  • Ability to move, swivel, and displace for optimal viewing position
  • Adapter and illumination included
StemiScope™ with optics and mount:
  • Eyepieces: 10x
  • Supplementary lens: 0.63x
  • Zoom magnifying range: 4.1 to 31.5x
  • Microscope adapter for angled view of specimen
  • Horizontal view and vertical drive and swivel mechanism
Swan-neck light guide:
  • 3.5mm fiber bundle
  • 50mm long swan-neck light guide
  • 750mm long flexible light guide
  • High light intensity
  • Adapter for the KL200 (760317) included
Specifications

Specifications

Standard Buffer Tray VN
Specimen stages B, F, and K
Standard Buffer Tray VN
Standard Buffer Tray VN
Videos
SDS
Documents
Certificates