missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HM13 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17121134 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17121134 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17121134 Supplier Invitrogen™ Supplier No. PA5113271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HM13 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Catalyzes intramembrane proteolysis of some signal peptides after they have been cleaved from a preprotein, resulting in the release of the fragment from the ER membrane into the cytoplasm. Required to generate lymphocyte cell surface (HLA-E) epitopes derived from MHC class I signal peptides. May play a role in graft rejection. May be necessary for the removal of the signal peptide that remains attached to the hepatitis C virus core protein after the initial proteolytic processing of the polyprotein. Involved in the intramembrane cleavage of the integral membrane protein PSEN1. Cleaves the integral membrane protein XBP1 isoform 1 in a DERL1/RNF139- dependent manner.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HM13
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene HM13
Gene Accession No. Q8TCT9, Q9D8V0
Gene Alias 1200006O09Rik; 4930443L17Rik; 5031424B04Rik; AV020344; cb228; H13; H-13; hIMP1; histocompatibility (minor) 13; histocompatibility 13; histocompatibility minor 13; HM13; hm13 protein; IMP1; IMP-1; IMPAS; IMPAS-1; intramembrane aspartyl protease; intramembrane protease 1; minor histocompatibility antigen 13; minor histocompatibility antigen H13; MSTP086; presenilin-like aspartyl protease; presenilin-like protein 3; PSENL3; PSL3; Signal peptide peptidase; signal peptide peptidase beta; signal peptide peptidase like 1; SPP; SPPL1; Unknown (protein for MGC:134054); wu:fc11a06; zgc:56660; zgc:86862
Gene Symbols H13, HM13
Host Species Rabbit
Immunogen Recombinant Human Minor histocompatibility antigen H13 protein (336-377AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14950, 81502
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.