missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMBS Antibody (OTI3F4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70894
This item is not returnable.
View return policy
Description
HMBS Monoclonal specifically detects HMBS in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HMBS | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| hydroxymethylbilane synthasePre-uroporphyrinogen synthase, PBGDPBG-D, PORC, porphobilinogen deaminase, porphyria, acute; Chester type, UPSEC 2.5.1.61, uroporphyrinogen I synthase, uroporphyrinogen I synthetase | |
| Mouse | |
| 39.1 kDa | |
| 100 μg | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI3F4 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| HMBS | |
| Full length human recombinant protein of human HMBS (NP_000181) produced in HEK293T cell. | |
| Affinity Purified | |
| Diabetes Research | |
| 3145 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction