Learn More
HMBS Monoclonal Antibody (OTI5B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA802690S
Encompass_Preferred
 View more versions of this product
Product Code. 17501333
  • € 242.00 / 30µL
Description
HMBS Monoclonal antibody specifically detects HMBS in Human samples. It is validated for Western Blot
Specifications
Specifications
HMBS
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P00480
Mouse
Affinity chromatography
RUO
5009
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5B11
Unconjugated
OTC
alternative name: porphobilinogen deaminase; HEM3; hemC; HMBS; Hydroxymethylbilane synthase; PBG D; PBGD; PBG-D; PORC; Porphobilinogen deaminase; porphyria, acute; Chester type; pre-uroporphyrinogen synthase; similar to Porphobilinogen deaminase (Hydroxymethylbilane synthase) (HMBS) (Pre-uroporphyrinogen synthase) (PBG-D); T25658; Ups; uroporphyrinogen I synthase; uroporphyrinogen I synthetase; URO-S; Uros1
Human recombinant protein fragment corresponding to amino acids 33-354 of human OTC produced in E.coli.