Learn More
HMBS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14797-1-AP-20UL
 View more versions of this product
Product Code. 16803624
  • € 145.00 / 20µL
Description
PBGD (porphobilinogen deaminase), also designated hydroxymethylbilane synthase, is a cytoplasmic enzyme found in the heme synthesis pathway. PBGD belongs to the HMBS (hydroxymethylbilane synthase) family. Deficiency of PBGD causes errors in pyrrole metabolism, which in turn leads to an inherited autosomal disorder called acute intermittent porphyria (AIP). AIP is characterized by acute attacks of neurological dysfunctions with hypertension, tachycardia, peripheral neurologic disturbances, abdominal pain and excessive amounts of aminolevulinic acid and porphobilinogen in the urine.
Specifications
Specifications
HMBS
Polyclonal
Unconjugated
HMBS
HMBS, hydroxymethylbilane synthase, PBG D, PBGD, Porphobilinogen deaminase, Pre uroporphyrinogen synthase, UPS
Rabbit
Antigen Affinity Chromatography
RUO
15288, 25709, 3145
-20°C
Liquid
Immunocytochemistry, Western Blot, Immunoprecipitation, Immunofluorescence
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P08397, P19356, P22907
Hmbs
HMBS Fusion Protein Ag6509
20 μL
Primary
Mouse, Rat, Human