missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97735PE
This item is not returnable.
View return policy
Description
HMG20A Polyclonal antibody specifically detects HMG20A in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| HMG20A | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| FLJ10739, high-mobility group 20A, HMG box domain containing 1, HMG box-containing protein 20A, HMG domain-containing protein 1, HMGX1HMG domain-containing protein HMGX1, HMGXB1high mobility group protein 20A | |
| Produced in rabbits immunized with E. coli-derived Human HMG20A fragment. | |
| 0.1 mL | |
| Chromatin Research, Stem Cell Markers | |
| 10363 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| PE | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction