Learn More
Invitrogen™ HMG20A Monoclonal Antibody (OTI2H4)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA526833
Product Code. 15956424
  • € 588.00 / 100µL
Description
HMG20A Monoclonal Antibody for Western Blot, IHC (P)
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HMG20A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NP66
HMG20A
Human recombinant protein fragment corresponding to amino acids 1-281 of HMG20A produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI2H4
Unconjugated
HMG20A
1200004E06Rik; 5730490E10Rik; FLJ10739; high mobility group 20A; high mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1; HMG20A; HMGX1; HMGXB1; Ibraf; Inhibitor of BRAF35; RGD1564760
Mouse
Affinity Chromatography
RUO
10363
-20°C, Avoid Freeze/Thaw Cycles