missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Monoclonal Antibody (OTI5E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807041
This item is not returnable.
View return policy
Description
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.Specifications
| HMG20A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NP66 | |
| HMG20A | |
| Human recombit protein fragment corresponding to amino acids 1-281 of human HMG20A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5E3 | |
| Unconjugated | |
| HMG20A | |
| HMGX1, HMGXB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10363 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction