missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG20A Monoclonal Antibody (OTI5E3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807041S
This item is not returnable.
View return policy
Description
HMG20A Monoclonal antibody specifically detects HMG20A in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HMG20A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99966 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4435 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5E3 | |
| Unconjugated | |
| CITED1 | |
| 1200004E06Rik; 5730490E10Rik; FLJ10739; high mobility group 20A; high mobility group protein 20A; HMG box domain containing 1; HMG box-containing protein 20A; HMG domain-containing protein 1; HMG domain-containing protein HMGX1; HMG20A; HMGX1; HMGXB1; Ibraf; Inhibitor of BRAF35; RGD1564760 | |
| Full length human recombinant protein of human CITED1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction