Learn More
HMG20A Mouse anti-Human, Clone: OTI5G3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807001
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Plays a role in neuronal differentiation as chromatin-associated protein. Acts as inhibitor of HMG20B. Overcomes the repressive effects of the neuronal silencer REST and induces the activation of neuronal-specific genes. Involved in the recruitment of the histone methyltransferase KMT2A/MLL1 and consequent increased methylation of histone H3 lysine 4.Specifications
| HMG20A | |
| Monoclonal | |
| Unconjugated | |
| HMG20A | |
| HMGX1, HMGXB1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10363 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI5G3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9NP66 | |
| HMG20A | |
| Human recombit protein fragment corresponding to amino acids 1-281 of human HMG20A produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.